Merck
  • Overexpression of BST2 is associated with nodal metastasis and poorer prognosis in oral cavity cancer.
Overexpression of BST2 is associated with nodal metastasis and poorer prognosis in oral cavity cancer.
The Laryngoscope (2014-04-08)
Ku-Hao Fang, Huang-Kai Kao, Lang-Ming Chi, Ying Liang, Shiau-Chin Liu, Chuen Hseuh, Chun-Ta Liao, Tzu-Chen Yen, Jau-Song Yu, Kai-Ping Chang
ABSTRACT
Bone marrow stromal cell antigen 2 (BST2) was one of the proteins that were found to be related to tumor metastasis in our previous proteomic study. Now we examine its clinical role on the oral cavity squamous cell carcinoma (OSCC). Individual retrospective cohort study and basic research. Immunohistochemical analysis, Western blotting, and quantitative real-time polymerase chain reaction were used to demonstrate the expression levels of BST2 on 159 OSCC tumors. RNA interference was utilized for cell migration and proliferation study in vitro. BST2 expression was significantly higher in OSCC cells of metastatic lymph nodes and primary tumor cells, compared to adjacent normal epithelia. Higher BST2 expression was associated with positive N stage, advanced overall stage, perineural invasion, and tumor depth (P = .049, .015, .021, and .010, respectively). OSCC patients with higher BST2 expression had poorer prognosis for disease-specific and disease-free survival (P = .009 and .001, respectively). Multivariate analyses also demonstrated that higher BST2 expression is an independent prognostic factor of disease-specific and disease-free survival (P = .047 and .013, respectively). In vitro suppression of BST2 expression in OEC-M1 cells showed that BST2 contributes to tumor migration of OSCC cells. The findings in this study indicate that BST2 expression in OSCC tumors is an independent prognostic factor of patient survival and associated with tumor metastasis.
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-BST2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 95%