Merck
  • Anti-inflammatory actions of herbal formula Gyejibokryeong-hwan regulated by inhibiting chemokine production and STAT1 activation in HaCaT cells.
Anti-inflammatory actions of herbal formula Gyejibokryeong-hwan regulated by inhibiting chemokine production and STAT1 activation in HaCaT cells.
Biological & pharmaceutical bulletin (2015-03-12)
Soo-Jin Jeong, Hye-Sun Lim, Chang-Seob Seo, Seong-Eun Jin, Sae-Rom Yoo, Nari Lee, Hyeun-Kyoo Shin
ABSTRACT
Gyejibokryeong-hwan (GJBRH; Keishi-bukuryo-gan in Japan and Guizhi Fuling Wan in China) is a traditional herbal formula comprising five medicinal herbs and is used to treat climacteric syndrome. GJBRH has been shown to exhibit biological activity against diabetes, diabetic nephropathy, atherosclerosis, ischemia, and cancer. However, there is no scientific evidence of its activities against skin inflammation, including atopic dermatitis. We used the HaCaT human keratinocyte cell line to investigate the effects of GJBRH on skin inflammation. No significant cytotoxicity was observed in cells treated with GJBRH up to a concentration of 1000 µg/mL. Exposure to the proinflammatory cytokines tumor necrosis factor-alpha (TNF-α) and interferon-gamma (IFN-γ) significantly increased HaCaT cell production of the following chemokines: macrophage-derived chemokine (MDC)/CCL22; regulated on activation, normal T-cell expressed and secreted (RANTES)/CCL5; and interleukin-8 (IL-8). In contrast, GJBRH significantly reduced the production of MDC, RANTES, and IL-8 compared with control cells simulated with TNF-α and IFN-γ. Consistently, GJBRH suppressed the mRNA expression of MDC, RANTES, and IL-8 in TNF-α and IFN-γ-treated cells. Treatment with GJBRH markedly inhibited phosphorylation of signal transducer and activator of transcription 1 (STAT1) in HaCaT cells stimulated with TNF-α and IFN-γ. Our findings indicate that GJBRH impairs TNF-α and IFN-γ-mediated inflammatory chemokine production and STAT1 phosphorylation in keratinocytes. We suggest that GJBRH may be a potent therapeutic agent for inflammatory skin disorders.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
trans-Cinnamaldehyde, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%
Sigma-Aldrich
trans-Cinnamaldehyde, FCC, FG
Sigma-Aldrich
Albiflorin, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels