Merck
  • Identification of 2-aminothiazolobenzazepine metabolites in human, rat, dog, and monkey microsomes by ion-molecule reactions in linear quadrupole ion trap mass spectrometry.
Identification of 2-aminothiazolobenzazepine metabolites in human, rat, dog, and monkey microsomes by ion-molecule reactions in linear quadrupole ion trap mass spectrometry.
Drug metabolism and disposition: the biological fate of chemicals (2014-12-31)
Minli Zhang, Ryan Eismin, Hilkka Kenttämaa, Hui Xiong, Ye Wu, Doug Burdette, Rebecca Urbanek
ABSTRACT
2-Aminothiazolobenzazepine (2-ATBA), 7-[(1-methyl-1H-pyrazol-4-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,3]thiazolo[4,5-h][3]benzazepin-2-amine, is a D2 partial agonist that has demonstrated antipsychotic effects in a rodent in vivo efficacy model. The metabolite profile showed that 2-ATBA is mainly metabolized by oxidation. However, identification of the oxidation site(s) in the 2-aminothiazole group presents a challenge for the traditional metabolite identification methods such as liquid chromatography/mass spectrometry and NMR due to the lack of unique tandem mass spectrometry fragmentation patterns for ions with the 2-aminothiazole group oxidized at different sites and the lack of stability for purification or reference standard synthesis. We describe the characterization of the oxidized heteroatoms of the 2-aminothiazole group via gas-phase ion-molecule reactions (GPIMR) in a modified linear quadrupole ion trap mass spectrometer. The GPIMR reagents used were dimethyl disulfide, tert-butyl peroxide, and tri(dimethylamino)borane. Each reagent was introduced into the ion trap through the helium line and was allowed to react with the protonated metabolites. The ionic ion-molecule reaction products and their fragmentation profiles were compared with the profiles of the ionic ion-molecule reaction products of protonated reference compounds that had specific heteroatom functionalities. The oxidized 2-aminothiazole metabolite of 2-ATBA showed a similar GPIMR profile to that of the reference compounds with a tertiary N-oxide functionality and distinct from the profiles of the reference compounds with N-aryl hydroxylamine, nitroso, or pyridine N-oxide functionalities. This study demonstrates the feasibility of fingerprinting the chemical nature of oxidized nitrogen functional groups via GPIMR profiling for metabolite structure elucidation.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%