Merck
  • Breaking the sensitivity limitations of cytochrome P450 oxidation product: dansyl chloride derivatisation of 4-OH mephenytoin, a CYP2C19 metabolite and its application to in vitro CYP inhibition assay.
Breaking the sensitivity limitations of cytochrome P450 oxidation product: dansyl chloride derivatisation of 4-OH mephenytoin, a CYP2C19 metabolite and its application to in vitro CYP inhibition assay.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-24)
V Vijayabhaskar, Pratima Srivastava, Sriram Rajagopal
ABSTRACT
A rapid selective and sensitive liquid chromatography/tandem mass spectrometry (LC-MS/MS) method was developed for the quantitative determination of derivatised cytochrome P450-2C19 oxidation product (dansyl-4-OH mephenytoin) and its underivatised form (4-OH mephenytoin). Samples were anaysed on C18 column (Waters Xbridge, 50 mm×4.6 mm, 3.5 μm particle size) with the mobile phase consisting of 0.1% formic acid in water and 0.1% formic acid in acetonitrile. A gradient method with a short run time of 2.5 min and 3.5 min was developed for the analysis of dansyl-4-OH mephenytoin and 4-OH mephenytoin, respectively. The standard curve was linear (r(2)=0.9972 for 4-OH mephenytoin; r(2)=0.9946 for dansyl-4-OH mephenytoin) over the concentration range of 0.16 to 40 ng/mL for both derivatised and underivatised forms. The CV (%) and relative error (RE) for inter and intraassay at three QC levels for dansyl-4-OH mephenytoin was 0.97-5.85% and -9.80 to 2.51%, respectively. Whereas, for 4-OH mephenytoin the CV (%) and RE (%) at three QC levels was 0.82-3.47% and -6.69 to -0.01%, respectively. The developed method was validated for various parameters such as linearity, precision & accuracy, extraction recovery, matrix effect, autosampler stability and was proved to be consistent across three QC levels with overall CV (%) less than 15. Dansylation helped in increasing the sensitivity of hydroxy mephenytoin by 100-200 fold. Given the simplicity involved in derivatisation process, we believe that this novel methodology will change the current approaches used for the enhancing the detection sensitivity of 4-OH mephenytoin.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Adenine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenine, ≥99%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Supelco
Dansyl chloride, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard