Merck
Crystals of Na(+)/K(+)-ATPase with bound cisplatin.
Biochemical pharmacology (2014-09-10)
Miroslav Huliciak, Linda Reinhard, Mette Laursen, Natalya Fedosova, Poul Nissen, Martin Kubala
ABSTRACT
Cisplatin is the most widely used chemotherapeutics for cancer treatment, however, its administration is connected to inevitable adverse effects. Previous studies suggested that cisplatin is able to inhibit Na(+)/K(+)-ATPase (NKA), the enzyme responsible for maintaining electrochemical potential and sodium gradient across the plasma membrane. Here we report a crystallographic analysis of cisplatin bound to NKA in the ouabain bound E2P form. Despite a moderate resolution (7.4 Å and 7.9 Å), the anomalous scattering from platinum and a model representation from a recently published structure enabled localization of seven cisplatin binding sites by anomalous difference Fourier maps. Comparison with NKA structures in the E1P conformation suggested two possible inhibitory mechanisms for cisplatin. Binding to Met151 can block the N-terminal pathway for transported cations, while binding to Met171 can hinder the interaction of cytoplasmic domains during the catalytic cycle.
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Dimethyl ether, ≥99.9%
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%