Merck
  • Bioassay-Guided Chromatographic Isolation and Identification of Antibacterial Compounds from Artemisia annua L. That Inhibit Clostridium perfringens Growth.
Bioassay-Guided Chromatographic Isolation and Identification of Antibacterial Compounds from Artemisia annua L. That Inhibit Clostridium perfringens Growth.
Journal of AOAC International (2015-04-24)
Elise Ivarsen, Xavier C Fretté, Kathrine B Christensen, Lars P Christensen, Ricarda M Engberg, Kai Grevsen, Anders Kjaer
ABSTRACT
Clostridium perfringens is the causative agent of necrotic enteritis leading to significant losses in the poultry industry. Dichloromethane and n-hexane extracts of aerial parts of Artemisia annua (Asteraceae) exhibited activity against C. perfringens with minimum inhibitory concentrations (MIC) of 185 and 270 μg/mL, respectively. Bioassay-guided fractionation of the extracts gave several active fractions (MIC between 75 and 600 μg/mL). Investigations of the most active fractions resulted in the isolation and characterization of the polyacetylene ponticaepoxide (MIC between 100 and 200 μg/mL) and (+)-threo-(5E)-trideca-1,5-dien-7,9,11-triyne-3,4-diol (MIC between 400 and 800 μg/mL), the flavonols chrysosplenol D (MIC between 200 and 400 μg/mL) and casticin (slight inhibition at 800 μg/mL), and 2,4-dihydroxy-6-methoxyacetophenone (slight inhibition at 800 μg/mL). Also, the coumarin scopoletin and the sesquiterpene lactone artemisinin were isolated from active fractions but showed no inhibition of C. perfringens growth at 800 and 2000 μg/mL, respectively. Fractions containing essential oil components with camphor constituting >60% did not show inhibition of C. perfringens up to 1600 μg/mL. Extracts and some active fractions showed higher antibacterial effect than individual bioactive compounds, suggesting that synergistic effects may underlie the observed antibacterial effect. The present study adds new valuable information on the antibacterial effect of A. annua against C. perfringens.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent