Merck
  • One-pot enzymatic conversion of carbon dioxide and utilization for improved microbial growth.
One-pot enzymatic conversion of carbon dioxide and utilization for improved microbial growth.
Environmental science & technology (2015-03-31)
Sung-Gil Hong, Hancheol Jeon, Han Sol Kim, Seung-Hyun Jun, EonSeon Jin, Jungbae Kim
ABSTRACT
We developed a process for one-pot CO2 conversion and utilization based on simple conversion of CO2 to bicarbonate at ambient temperature with no energy input, by using the cross-linking-based composites of carboxylated polyaniline nanofibers (cPANFs) and carbonic anhydrase. Carbonic anhydrase was immobilized on cPANFs via the approach of magnetically separable enzyme precipitate coatings (Mag-EPC), which consists of covalent enzyme attachment, enzyme precipitation, and cross-linking with amine-functionalized magnetic nanoparticles. Mag-EPC showed a half-life of 236 days under shaking, even resistance to 70% ethanol sterilization, and recyclability via facile magnetic separation. For one-pot CO2 conversion and utilization, Mag-EPC was used to accelerate the growth of microalga by supplying bicarbonate from CO2, representing 1.8-fold increase of cell concentration when compared to the control sample. After two repeated uses via simple magnetic separation, the cell concentration with Mag-EPC was maintained as high as the first cycle. This one-pot CO2 conversion and utilization is an alternative as well as complementary process to adsorption-based CO2 capture and storage as an environmentally friendly approach, demanding no energy input based on the effective action of the stabilized enzyme system.
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3-Aminobenzamide, ≥99% (TLC)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM