Merck
  • Highly lipophilic 3-epi-betulinic acid derivatives as potent and selective TGR5 agonists with improved cellular efficacy.
Highly lipophilic 3-epi-betulinic acid derivatives as potent and selective TGR5 agonists with improved cellular efficacy.
Acta pharmacologica Sinica (2014-10-07)
Xiao-yin Wang, Shu-yong Zhang, Jing Li, Hua-nan Liu, Xin Xie, Fa-jun Nan
ABSTRACT
TGR5 is a G protein-coupled receptor that is expressed in intestinal L-cells and stimulates glucagon-like peptide 1 (GLP-1) secretion. TGR5 may represent a novel target for the treatment of metabolic disorder. Here, we sought to design and synthesize a series of TGR5 agonists derived from the natural product betulinic acid. A series of betulinic acid derivatives were designed and synthesized. A cAMP assay was established using a HEK293 cell line expressing human TGR5. Luciferase reporter assay was established using HEK293 cells transfected with plasmids encoding human FXR and luciferase reporter. A human intestinal L-cell line NCI-H716 was used to evaluate the effects of the betulinic acid derivatives on GLP-1 secretion in vitro. Biological data revealed that the 3-α-OH triterpenoids consistently show increased potency for TGR5 compared to their 3-β-OH epimers. 3-OH esterification increased the lipophilicity and TGR5 activity of 3-α betulinic derivatives and enhanced the activity differences between 3-α and 3-β derivatives. The 3-α-acyloxy betulinic acids also exhibited a significant dose-dependent GLP-1 secretion effect. This study demonstrates that highly lipophilic 3-epi-betulinic acid derivatives can be potent and selective TGR5 agonists with improved cellular efficacy, and our research here provides a new strategy for the design and development of potent TGR5 agonists.
Product Description
Lithocholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lithocholic acid, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard