Merck
  • Host-habitat location by the parasitoid, Nasonia vitripennis Walker (Hymenoptera: Pteromalidae).
Host-habitat location by the parasitoid, Nasonia vitripennis Walker (Hymenoptera: Pteromalidae).
Journal of forensic sciences (2013-08-29)
Christine Frederickx, Jessica Dekeirsschieter, Francois J Verheggen, Eric Haubruge
ABSTRACT
This study investigated the role of odorant cues used during host-habitat location by the generalist parasitoid, Nasonia vitripennis Walker. Nasonia vitripennis is a common parasitoid of Dipteran pupae found in association with decaying carrion. Behavioral assays were used to investigate the host-habitat searching behavior under different scenarios. First, we demonstrated N. vitripennis to be significantly attracted toward odorant cues associated with decaying meat. The biological activity of nine of the volatile molecules constituting the odor of decaying meat was tested on the searching behavior of parasitoid females through two complementary chemoecological approaches: electroantennography (EAG) and olfactometry bioassays. Butanoic acid and butan-1-ol elicited high olfactory responses, but no attraction was induced by these two chemicals. Behavioral assays showed that, among the VOCs tested, methyldisulfanylmethane (DMDS) was the only volatile chemical to induce attraction in N. vitripennis.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Dimethyl disulfide, ≥99.0%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl disulfide, ≥99%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Dimethyl disulfide, ≥98%, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
Dimethyl disulfide, analytical standard
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%