Merck
  • Metastasis-associated in colon cancer 1 is an independent prognostic biomarker for survival in Klatskin tumor patients.
Metastasis-associated in colon cancer 1 is an independent prognostic biomarker for survival in Klatskin tumor patients.
Hepatology (Baltimore, Md.) (2015-05-09)
Andri Lederer, Pia Herrmann, Daniel Seehofer, Manfred Dietel, Johann Pratschke, Peter Schlag, Ulrike Stein
ABSTRACT
Curative treatment of intrahepatic cholangiocarcinoma (ICC) and hilar cholangiocarcinoma (Klatskin tumors) is limited to surgical resection or orthotopic liver transplantation. However, not all patients benefit from a surgical approach and suffer from early tumor recurrence. Response to chemotherapy is generally poor and, until today, no targeted therapy could be established. Metastasis-associated in colon cancer 1 (MACC1) is a recently discovered regulator of the hepatocyte growth factor (HGF)/Met/mitogen-activated protein kinase pathway, which induces proliferation, migration, and invasion in cell culture, as well as metastasis in mice. MACC1 expression shows a significant correlation with Met expression in colon cancer tissue and is highly prognostic for occurrence of distant metastasis and survival in colon cancer patients. Thus, we aimed to measure the expression of MACC1, Met, and HGF messenger RNA in microdissected tumor tissue and corresponding normal liver tissue of 156 patients with Klatskin tumors (n = 76) and ICC (n = 80) using real-time quantitative reverse-transcriptase polymerase chain reaction. We used immunohistochemical staining to validate the results. MACC1 expression in tumor tissue of both tumor entities was significantly higher than in corresponding normal liver tissue (P < 0.001). Klatskin tumor patients with a history of tumor recurrence had significantly higher MACC1 expression than those without tumor recurrence (P = 0.005). Uni- und multivariate survival analysis showed that Klatskin tumor patients with high MACC1 had a significantly shorter overall (OS) and disease-free survival (DFS; P = 0.001 and P < 0.001, respectively). The multivariate analysis confirmed MACC1 to be an independent factor for overall survival in Klatskin tumor patients (hazard ratio: 2.777; 95% confidence interval: 1.389-5.555; P = 0.004). Our study identified MACC1 as a highly prognostic biomarker for OS and DFS in Klatskin tumor patients. MACC1 expression could become an important diagnostic tool and might be a candidate for targeted therapy.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O