Merck
  • Crystal structure of Get4-Get5 complex and its interactions with Sgt2, Get3, and Ydj1.
Crystal structure of Get4-Get5 complex and its interactions with Sgt2, Get3, and Ydj1.
The Journal of biological chemistry (2010-01-29)
Yi-Wei Chang, Yu-Chien Chuang, Yu-Chi Ho, Ming-Yuan Cheng, Yuh-Ju Sun, Chwan-Deng Hsiao, Chung Wang
ABSTRACT
Get3, Get4, and Get5 in Saccharomyces cerevisiae participate in the insertion of tail-anchored proteins into the endoplasmic reticulum membrane. We elucidated the interaction between Get4 and Get5 and investigated their interaction with Get3 and a tetratricopeptide repeat-containing protein, Sgt2. Based on co-immunoprecipitation and crystallographic studies, Get4 and Get5 formed a tight complex, suggesting that they constitute subunits of a larger complex. In contrast, although Get3 interacted physically with the Get4-Get5 complex, low amounts of Get3 co-precipitated with Get5, implying a transient interaction between Get3 and Get4-Get5. Sgt2 also interacted with Get5, although the amount of Sgt2 that co-precipitated with Get5 varied. Moreover, GET3, GET4, and GET5 interacted genetically with molecular chaperone YDJ1, suggesting that chaperones might also be involved in the insertion of tail-anchored proteins.