Merck
  • Use of a standardized JaCVAM in vivo rat comet assay protocol to assess the genotoxicity of three coded test compounds; ampicillin trihydrate, 1,2-dimethylhydrazine dihydrochloride, and N-nitrosodimethylamine.
Use of a standardized JaCVAM in vivo rat comet assay protocol to assess the genotoxicity of three coded test compounds; ampicillin trihydrate, 1,2-dimethylhydrazine dihydrochloride, and N-nitrosodimethylamine.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-07-28)
J P McNamee, P V Bellier
ABSTRACT
As part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiative international validation study of the in vivo rat alkaline comet assay (comet assay), our laboratory examined ampicillin trihydrate (AMP), 1,2-dimethylhydrazine dihydrochloride (DMH), and N-nitrosodimethylamine (NDA) using a standard comet assay validation protocol (v14.2) developed by the JaCVAM validation management team (VMT). Coded samples were received by our laboratory along with basic MSDS information. Solubility analysis and range-finding experiments of the coded test compounds were conducted for dose selection. Animal dosing schedules, the comet assay processing and analysis, and statistical analysis were conducted in accordance with the standard protocol. Based upon our blinded evaluation, AMP was not found to exhibit evidence of genotoxicity in either the rat liver or stomach. However, both NDA and DMH were observed to cause a significant increase in % tail DNA in the rat liver at all dose levels tested. While acute hepatoxicity was observed for these compounds in the high dose group, in the investigators opinion there were a sufficient number of consistently damaged/measurable cells at the medium and low dose groups to judge these compounds as genotoxic. There was no evidence of genotoxicity from either NDA or DMH in the rat stomach. In conclusion, our laboratory observed increased DNA damage from two blinded test compounds in rat liver (later identified as genotoxic carcinogens), while no evidence of genotoxicity was observed for the third blinded test compound (later identified as a non-genotoxic, non-carcinogen). This data supports the use of a standardized protocol of the in vivo comet assay as a cost-effective alternative genotoxicity assay for regulatory testing purposes.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
5,5-Dimethylhydantoin, 97%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture