Merck
  • Antioxidative and immunological activities of ophiopogon polysaccharide liposome from the root of Ophiopogon japonicus.
Antioxidative and immunological activities of ophiopogon polysaccharide liposome from the root of Ophiopogon japonicus.
Carbohydrate polymers (2015-10-11)
Yunpeng Fan, Xia Ma, Lin Ma, Jing Zhang, Weimin Zhang, Xiaoping Song
ABSTRACT
The antioxidative and immunoregulatory activities of ophiopogon polysaccharide liposome (OPL) were investigated. The results demonstrated that the radical-scavenging activity increased along with the increasing of OPL concentrations, and the scavenging capability of OPL was slightly stronger than ophiopogon polysaccharide (OP) in vitro. In vivo, OPL at high and medium doses could significantly improve the levels of SOD and GSH-Px, and decrease the levels of MPO, XOD and MDA compared with OP. In addition, OPL could not only significantly enhance the splenocyte proliferation and the proportion of CD4(+) and CD8(+) T cells, but also improve the levels of cytokines, antigen-specific antibody titers and immune organ index in the mice immunized with OVA compared with OP. These results indicated that the antioxidative and immunoregulatory activities of OP were significantly enhanced after encapsulated with liposome. Therefore, OPL would be expected to exploit into a new-type of functional food as both medicine and food.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%