Merck
  • Clopidogrel reduces apoptosis and promotes proliferation of human vascular endothelial cells induced by palmitic acid via suppression of the long non-coding RNA HIF1A-AS1 in vitro.
Clopidogrel reduces apoptosis and promotes proliferation of human vascular endothelial cells induced by palmitic acid via suppression of the long non-coding RNA HIF1A-AS1 in vitro.
Molecular and cellular biochemistry (2015-03-13)
Jing Wang, Lingqiang Chen, Hongfei Li, Jin Yang, Zhiqiang Gong, Bing Wang, Xueling Zhao
ABSTRACT
Cardiovascular disease (CVD) is recognized as a major and increasing health problem affected older subjects in China, and clopidogrel has been widely used for treatment of CVD patients such as atherosclerosis, myocardial infarction, and myocardial ischaemia-reperfusion damage. However, the molecular mechanisms of clopidogrel for treatment of CVD are only partially understood. This study investigated the effects of clopidogrel on palmitic acid-induced damage of human vascular endothelial cells (HUVECs), and the molecular mechanisms of LncRNA HIF1A-AS1 in regulating the proliferation and apoptosis of HUVECs in vitro. We firstly established a damage model of HUVECs through palmitic acid (PA) treatment. And the effect of clopidogrel reducing PA-induced apoptosis of HUVECs was observed by the flow cytometric measurement. To further understand the molecular mechanism of clopidogrel rescues PA-induced apoptosis, we used human LncRNA PCR array to compare the LncRNA expression profile difference between clopidogrel-treated cells and control cells. The expression of LncRNA HIF 1 alpha-antisense RNA 1 (HIF1A-AS1) was significantly altered in clopidogrel-treated cells. We further proved that suppression of HIF1A-AS1 by siRNA reduce PA-induced apoptosis and promote proliferation of HUVECs. Furthermore, we also demonstrated inhibition apoptosis effect by HIF1A-AS1 is related to mitochondrial apoptosis pathway. Hence, our results suggest that clopidogrel rescues apoptosis and promotes proliferation of PA-induced damage model of HUVECs through inhibiting the mediator LncRNA HIF1A-AS1. These findings indicate that LncRNA HIF1A-AS1 may play an important role in the pathogenesis of CVD, and provide a novel molecular mechanism of clopidogrel for treatment of CVD.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder