  • Search Results
  • Comparative proteomic analysis of Taurine, Indicine, and crossbred (Bos taurus × Bos indicus) bull spermatozoa for identification of proteins related to sperm malfunctions and subfertility in crossbred bulls.
Comparative proteomic analysis of Taurine, Indicine, and crossbred (Bos taurus × Bos indicus) bull spermatozoa for identification of proteins related to sperm malfunctions and subfertility in crossbred bulls.
Theriogenology (2015-06-03)
Munchakkal Kather Muhammad Aslam, Arumugam Kumaresan, Shailendra Kumar Rajak, Md Tajmul, Tirtha Kumar Datta, Tushar Kumar Mohanty, Alagiri Srinivasan, Savita Yadav
ABSTRACT
Subfertility is one of the most common problems observed among Taurine × Indicine crossbred bulls in tropical countries; however, the etiology remain unknown in most of the cases. In present study, we compared the proteomic profile of spermatozoa from crossbred bulls (Bos taurus × Bos indicus) against their purebred parent lines (Holstein Friesian [Taurine] and Tharparkar [Indicine]) to find out alteration in expressions of proteins, if any. The proteomic profiles of freshly ejaculated spermatozoa from these breeds were compared by two-dimensional difference gel electrophoresis, and differentially expressed proteins were identified through mass spectrometry. It was observed that compared to Holstein Friesian, nine proteins were underexpressed and eight proteins were overexpressed (P < 0.05) in the spermatozoa of crossbred bulls. Similarly, four proteins were overexpressed and four proteins were underexpressed (P < 0.05) in the spermatozoa of crossbred bulls compared to Tharparkar bulls. In concurrent three breed comparison, 14 proteins were found to be differentially expressed (P < 0.05) between these breeds. From the findings of the study, it is apparent that the expression levels of several functionally significant proteins are either upregulated or downregulated in spermatozoa of crossbred bulls, which might be related to high incidence of subfertility in these bulls.
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)