Merck
Milk-clotting mechanism of Dregea sinensis Hemsl. protease.
Journal of dairy science (2015-10-28)
Yali Zhang, Hongyan Wang, Liang Tao, Ai-xiang Huang
ABSTRACT
Dregea sinensis Hemsl. is used as a milk coagulant to produce goat milk cakes in Yunnan, China. However, the composition of milk-clotting compounds and the related mechanism have not been reported. Crude protease was extracted from the stem, purified, and then separated with a Millipore ultrafiltration centrifuge tube. Cysteine protease (procerain B) was identified as the main milk-clotting protein through electrospray ionization mass spectrometry, and its molecular weight was 23.8 kDa. The protease can partially degrade α-casein (CN) and completely degrade β- and κ-CN, and κ-CN degradation resulted in milk clotting. The molecular weight and AA sequence of the peptide fractions were determined through matrix-assisted laser desorption/ionization time-of-flight mass spectrometry and a peptide sequencer, respectively. The enzyme cleaved κ-CN at Ala90-Gln91 and produced deputy κ-CN and caseinomacropeptide with molecular weights of 12 and 6.9 kDa, respectively. This cleavage site differed from the majority of chymosins cleaved at Phe105-Met106.
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)