  • Search Results
  • Comparative proteomic study of Edwardsiella tarda strains with different degrees of virulence.
Comparative proteomic study of Edwardsiella tarda strains with different degrees of virulence.
Journal of proteomics (2015-05-17)
Noemí Buján, Carolina Hernández-Haro, Lucía Monteoliva, Concha Gil, Beatriz Magariños
ABSTRACT
Edwardsiella tarda is an enteric opportunistic pathogen that causes a great loss in aquaculture. This species has been described as a phenotypical homogeneous group; in contrast, serological studies and molecular typing revealed a wide heterogeneity. In this work, a proteomic study of differential expression of a virulent isolate from turbot cultured in the Norwest of Spain in comparison with an avirulent collection strain was performed in order to recognize proteins involved in virulence. One hundred and three proteins that presented different abundance were successfully identified and classified into 11 functional categories according to their biological processes: amino acid, carbohydrate and lipid metabolism, tricarboxylic cycle, stress response and protein fate, protein synthesis, biogenesis of cellular components, cell rescue defence and virulence, cell membrane and transport, signal transduction and purine and pyrimidine metabolism. Twenty three protein spots detected only in turbot isolate were identified. It was shown that the same proteins appeared in different spots in the two isolates. Mass spectra obtained by MALDITOF/TOF of some of these proteins and DNA sequencing explained the changes as a result of different amino acid sequences. Several proteins related with the virulence of E. tarda (FliC, ArnA or FeSODI) were only detected in the turbot European isolate. This article is part of a Special Issue entitled: HUPO 2014.
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O