Merck
  • Nonalcoholic Fatty Liver Disease Is Independently Associated with Early Left Ventricular Diastolic Dysfunction in Patients with Type 2 Diabetes.
Nonalcoholic Fatty Liver Disease Is Independently Associated with Early Left Ventricular Diastolic Dysfunction in Patients with Type 2 Diabetes.
PloS one (2015-08-08)
Alessandro Mantovani, Matteo Pernigo, Corinna Bergamini, Stefano Bonapace, Paola Lipari, Isabella Pichiri, Lorenzo Bertolini, Filippo Valbusa, Enrico Barbieri, Giacomo Zoppini, Enzo Bonora, Giovanni Targher
ABSTRACT
Accumulating evidence suggests that nonalcoholic fatty liver disease (NAFLD) is associated with left ventricular diastolic dysfunction (LVDD) in nondiabetic individuals. To date, there are very limited data on this topic in patients with type 2 diabetes and it remains uncertain whether NAFLD is independently associated with the presence of LVDD in this patient population. We performed a liver ultrasonography and trans-thoracic echocardiography (with speckle-tracking strain analysis) in 222 (156 men and 66 women) consecutive type 2 diabetic outpatients with no previous history of ischemic heart disease, chronic heart failure, valvular diseases and known hepatic diseases. Binary logistic regression analysis was used to examine the association between NAFLD and the presence/severity of LVDD graded according to the current criteria of the American Society of Echocardiography, and to identify the variables that were independently associated with LVDD, which was included as the dependent variable. Patients with ultrasound-diagnosed NAFLD (n = 158; 71.2% of total) were more likely to be female, overweight/obese, and had longer diabetes duration, higher hemoglobin A1c and lower estimated glomerular filtration rate (eGFR) than those without NAFLD. Notably, they also had a remarkably greater prevalence of mild and/or moderate LVDD compared with those without NAFLD (71% vs. 33%; P<0.001). Age, hypertension, smoking, medication use, E/A ratio, LV volumes and mass were comparable between the two groups of patients. NAFLD was associated with a three-fold increased odds of mild and/or moderate LVDD after adjusting for age, sex, body mass index, hypertension, diabetes duration, hemoglobin A1c, eGFR, LV mass index and ejection fraction (adjusted-odds ratio 3.08, 95%CI 1.5-6.4, P = 0.003). In conclusion, NAFLD is independently associated with early LVDD in type 2 diabetic patients with preserved systolic function.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications