Merck
  • Novel Chiral Bifunctional L-Thiazoline-Thiourea Derivatives: Design and Application in Enantioselective Michael Reactions.
Novel Chiral Bifunctional L-Thiazoline-Thiourea Derivatives: Design and Application in Enantioselective Michael Reactions.
Chirality (2015-10-03)
Qi Lai, Yang Li, Zhiyong Gong, Qingwen Liu, Chiyu Wei, Zhiguang Song
ABSTRACT
Several novel chiral bifunctional L-thiazoline-thiourea derivatives were easily synthesized from commercially available L-cysteine in high yield. These catalysts were subsequently applied to the enantioselective Michael addition of acetylacetone to β-nitrostyrenes. The products with S configuration were obtained in 98% enantiomeric excess (ee) when the L-thiazoline-thiourea derivatives were used. A plausible transition state model is proposed to explain the observed enantioselectivities.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%