Merck
  • The NR2B antagonist, ifenprodil, corrects the l-DOPA-induced deficit of bilateral movement and reduces c-Fos expression in the subthalamic nucleus of hemiparkinsonian rats.
The NR2B antagonist, ifenprodil, corrects the l-DOPA-induced deficit of bilateral movement and reduces c-Fos expression in the subthalamic nucleus of hemiparkinsonian rats.
Neuroscience research (2015-02-24)
Masakazu Igarashi, Toshiya Habata, Hisanao Akita, Kazuko Noda, Masanori Ogata, Makoto Saji
ABSTRACT
The use of NR2B antagonists in Parkinsonism is still controversial. To examine their anti-parkinsonian effects, the NR2B antagonist, ifenprodil, and L-DOPA were administered together and separately in hemiparkinsonian rats (hemi-PD) that were subjected to a cylinder test. Recovery from hypoactivity was achieved by single administration of 3-7 mg/kg of L-DOPA; however, improvement in the deficit of bilateral forelimb use was not observed. When administered alone, ifenprodil had no anti-parkinsonian effects; however, combined administration of ifenprodil and 7 mg/kg of L-DOPA significantly reversed the deficit of bilateral forelimb use without adversely affecting the L-DOPA-induced improvement in motor activity. Next, in order to identify the brain area influenced by L-DOPA and ifenprodil, quantitative analysis of L-DOPA-induced c-Fos immunoreactivity was performed in various brain areas of hemi-PD following administration of L-dopa with and without ifenprodil. Among brain areas with robust c-Fos expression within the motor loop circuit in dopamine-depleted hemispheres, co-administered ifenprodil markedly attenuated L-DOPA-induced c-Fos expression in only the subthalamic nucleus (STN), suggesting that the STN is the primary target for the anti-parkinsonian action of NR2B antagonists.
Product Description
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Ethylene glycol 5 M solution