Merck
  • Curcumin attenuates ethanol-induced hepatic steatosis through modulating Nrf2/FXR signaling in hepatocytes.
Curcumin attenuates ethanol-induced hepatic steatosis through modulating Nrf2/FXR signaling in hepatocytes.
IUBMB life (2015-08-26)
Chunfeng Lu, Feng Zhang, Wenxuan Xu, Xiafei Wu, Naqi Lian, Huanhuan Jin, Qin Chen, Lianyun Chen, Jiangjuan Shao, Li Wu, Yin Lu, Shizhong Zheng
ABSTRACT
Alcoholic liver disease (ALD) is a common health problem worldwide, characterized by aberrant accumulation of lipid in hepatocytes. Inhibition of lipid accumulation has been well recognized as a promising strategy for ALD. Previous studies showed that curcumin has potential effect on ALD by regulating oxidative stress and ethanol metabolism. However, the effects of curcumin on lipid accumulation and its mechanism remain unclear. Recent researches have indicated that farnesoid X receptor (FXR) and nuclear factor (erythroid-derived 2)-like 2 (Nrf2) have excellent effects on reducing lipid deposition. This study demonstrated that curcumin alleviated ethanol-induced liver injury by ameliorating activities of serum marker enzymes and inflammation. Moreover, curcumin alleviated the symptom of hyperlipidemia and hepatic steatosis via modulating the expression of sterol regulatory element-binding protein-1c, fatty acid synthase, and peroxisome proliferator-activated receptor-alpha as well as the activity of carnitine palmitoyltransferase 1. Additionally, curcumin induced the expression of Nrf2 and FXR in liver, strongly implying close relationship between inhibitory effect of curcumin on hepatic steatosis and the above two genes. The following in vitro experiments further verified the protective effects of curcumin against hepatotoxicity and lipid accumulation in hepatocytes induced by ethanol. Gain- or loss-of-function analyses revealed Nrf2 and FXR mediated the effect of curcumin on lipid deposition in hepatocytes, and curcumin modulated the expression of FXR mediated by Nrf2. Collectively, we drew a conclusion that curcumin attenuated ALD by modulating lipid deposition in hepatocytes via a Nrf2/FXR activation-dependent mechanism. The findings make curcumin a potential agent for ALD and broaden the horizon of the molecular mechanism involved.
Sigma-Aldrich
GW4064, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nfe2l2
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent