Merck
  • Epithelial-mesenchymal transition induces similar metabolic alterations in two independent breast cancer cell lines.
Epithelial-mesenchymal transition induces similar metabolic alterations in two independent breast cancer cell lines.
Cancer letters (2015-04-29)
Yuvabharath Kondaveeti, Irene K Guttilla Reed, Bruce A White
ABSTRACT
Epithelial-mesenchymal transition (EMT) induces invasive properties in epithelial tumors and promotes metastasis. Although EMT-mediated cellular and molecular changes are well understood, very little is known about EMT-induced metabolic changes. HER2-positive BT-474 breast cancer cells were induced to undergo a stable EMT using mammosphere culture, as previously described by us for the ERα-positive MCF-7 breast cancer cells. Two epithelial breast cancer cell lines (BT-474 and MCF-7) were compared to their respective EMT-derived mesenchymal progeny (BT-474(EMT) and MCF-7(EMT)) for changes in metabolic pathways including glycolysis, glycogen metabolism, anabolic pathways and gluconeogenesis. Both EMT-derived cells displayed enhanced aerobic glycolysis along with the overexpression of specific glucose transporters, lactate dehydrogenase isoforms, monocarboxylate transporters and glycogen phosphorylase isoform. In contrast, both EMT-derived cells suppressed the expression of crucial enzymes in anabolic pathways and gluconeogenesis. STAT3, a transcription factor involved in tumor initiation and progression, plays a role in the EMT-related changes in the expression of specific enzymes and transporters. This study provides a broad overview of similar metabolic changes induced by EMT in two independent breast cancer cell lines. These metabolic changes may provide novel therapeutic targets for metastatic breast cancer.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)