Merck
  • Inclusion of homologous DNA in nuclease-mediated gene targeting facilitates a higher incidence of bi-allelically modified cells.
Inclusion of homologous DNA in nuclease-mediated gene targeting facilitates a higher incidence of bi-allelically modified cells.
Xenotransplantation (2015-09-19)
Benjamin P Beaton, Deug-Nam Kwon, Yun-Jung Choi, Jae-Hwan Kim, Melissa S Samuel, Joshua A Benne, Kevin D Wells, Kiho Lee, Jin-Hoi Kim, Randall S Prather
ABSTRACT
Recent advancements in gene editing techniques have increased in number and utility. These techniques are an attractive alternative to conventional gene targeting methods via homologous recombination due to the ease of use and the high efficiency of gene editing. We have previously produced cytidine monophosphate-N-acetylneuraminic acid hydroxylase (CMAH) knockout (KO) pigs in a Minnesota miniature pig genetic background. These pigs were generated using zinc-finger nucleases (ZFNs) in combination with donor DNA containing a total homology length of 1600 bp (800-bp homology on each arm). Our next aim was to introduce the targeted disruption of alpha-1,3-galactosyltransferase (GGTA1) in the CMAH KO genetic background and evaluate the effect of donor DNA homology length on meganuclease-mediated gene targeting. Zinc-finger nucleases from a previous CMAH KO experiment were used as a proof of concept to identify a correlation between the length of donor DNA homology and targeting efficiency. Based on those results, experiments were designed to use transcription activator-like effector nucleases (TALENs) to generate bi-allelically modified GGTA1 cells using donor DNAs carrying various lengths of homology. Donor DNA was designed to symmetrically flank the predicted cleavage sites in CMAH and GGTA1 for both ZFN and TALEN cleavage sites, respectively. For both genes, the length of total homology ranged from 60 to 1799 bp. Sialyltransferase gene expression profiles were evaluated in CMAH and GGTA1 double KO pig cells and were compared to wild-type and CMAH KO cells. Introduction of donor DNA with ZFNs demonstrated that small amounts of homology (60 bp) could facilitate homology-directed repair during ZFN-mediated targeting of CMAH; however, donor DNA with longer amounts of homology resulted in a higher frequency of homology-directed repair. For the GGTA1 KO experiments that used TALENs and donor DNA, donor DNA alone did not result in detectable bi-allelic conversion of GGTA1. As the length of donor DNA increased, the bi-allelic disruption of GGTA1 increased from 0.5% (TALENs alone, no donor DNA present) to a maximum of 3% (TALENs and donor DNA with total homology of 1799 bp). Inclusion of homologous donor DNA in TALEN-mediated gene targeting facilitated a higher incidence of bi-allelically modified cells. Using the generated cells, we were able to demonstrate the lack of GGTA1 expression and the decrease in gene expression sialyltransferase-related genes. The approach of using donor DNA in conjunction with a meganuclease can be used to increase the efficiency of gene targeting. The gene editing methods can be applied to other genes as well as other mammalian systems. Additionally, gene expression analysis further confirms that the CMAH/GGTA1 double KO pigs can be a valuable source for the study of pig-to-human xenotransplantation.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder