Merck
  • Compound 48/80, a Mast Cell Stimulator, Enhances Synthesis of IgE and IgG Induced by Intranasal Application of Ovalbumin in Mice.
Compound 48/80, a Mast Cell Stimulator, Enhances Synthesis of IgE and IgG Induced by Intranasal Application of Ovalbumin in Mice.
Biological & pharmaceutical bulletin (2015-12-04)
Nobuaki Matsui, Daisuke Ito, Yukari Takabatake, Eri Nashioka, Shingo Tada, Masato Kanagawa, Nobuyuki Fukuishi, Masaaki Akagi
ABSTRACT
Mast cells are well established effector cells of type I hypersensitivity reactions such as allergic rhinitis. However, recent studies have suggested that activated mast cells enhance local immunoglobulin E (IgE) synthesis in the nasal mucosa of allergic rhinitis patients. Therefore, we hypothesized that non-immunological mast cell activators may have the potential to enhance local IgE synthesis. Here, we examined the effect of compound 48/80 (C48/80), a mast cell activator, on IgE and immunoglobulin G (IgG) synthesis. Female Balb/c mice were intranasally administered a mixture of ovalbumin (OVA) (1-10 µg/nose) and C48/80 (1-100 µg/nose) on days 0, 7, 14 and 21 and on consecutive days from day 28 to day 42. Intranasal administration of C48/80 with OVA increased serum OVA-specific IgE and IgG. Double staining with fluorescent-labeled OVA and fluorescent-labeled IgE- or IgG-specific antibody demonstrated the presence of OVA-specific IgE- or IgG-producing cells in the nasal mucosa of sensitized mice. Moreover, intranasal administration of C48/80 with OVA increased the nasal mucosal interleukin (IL)-4 level and enhanced the OVA-induced symptom of sneezing. These results suggested that simultaneous activation of mast cells with antigen exposure enhances local IgE and IgG synthesis.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder