Merck
  • Experimental Evidence on the Formation of Ethene through Carbocations in Methanol Conversion over H-ZSM-5 Zeolite.
Experimental Evidence on the Formation of Ethene through Carbocations in Methanol Conversion over H-ZSM-5 Zeolite.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-17)
Chao Wang, Xianfeng Yi, Jun Xu, Guodong Qi, Pan Gao, Weiyu Wang, Yueying Chu, Qiang Wang, Ningdong Feng, Xiaolong Liu, Anmin Zheng, Feng Deng
ABSTRACT
The methanol to olefins conversion over zeolite catalysts is a commercialized process to produce light olefins like ethene and propene but its mechanism is not well understood. We herein investigated the formation of ethene in the methanol to olefins reaction over the H-ZSM-5 zeolite. Three types of ethylcyclopentenyl carbocations, that is, the 1-methyl-3-ethylcyclopentenyl, the 1,4-dimethyl-3-ethylcyclopentenyl, and the 1,5-dimethyl-3-ethylcyclopentenyl cation were unambiguously identified under working conditions by both solid-state and liquid-state NMR spectroscopy as well as GC-MS analysis. These carbocations were found to be well correlated to ethene and lower methylbenzenes (xylene and trimethylbenzene). An aromatics-based paring route provides rationale for the transformation of lower methylbenzenes to ethene through ethylcyclopentenyl cations as the key hydrocarbon-pool intermediates.