Merck
  • Cholesterol-Independent Suppression of Lymphocyte Activation, Autoimmunity, and Glomerulonephritis by Apolipoprotein A-I in Normocholesterolemic Lupus-Prone Mice.
Cholesterol-Independent Suppression of Lymphocyte Activation, Autoimmunity, and Glomerulonephritis by Apolipoprotein A-I in Normocholesterolemic Lupus-Prone Mice.
Journal of immunology (Baltimore, Md. : 1950) (2015-10-16)
Leland L Black, Roshni Srivastava, Trenton R Schoeb, Ray D Moore, Stephen Barnes, Janusz H Kabarowski
ABSTRACT
Apolipoprotein (Apo)A-I, the major lipid-binding protein of high-density lipoprotein, can prevent autoimmunity and suppress inflammation in hypercholesterolemic mice by attenuating lymphocyte cholesterol accumulation and removing tissue-oxidized lipids. However, whether ApoA-I mediates immune-suppressive or anti-inflammatory effects under normocholesterolemic conditions and the mechanisms involved remain unresolved. We transferred bone marrow from systemic lupus erythematosus (SLE)-prone Sle123 mice into normal, ApoA-I-knockout (ApoA-I(-/-)) and ApoA-I-transgenic (ApoA-I(tg)) mice. Increased ApoA-I in ApoA-I(tg) mice suppressed CD4(+) T and B cell activation without changing lymphocyte cholesterol levels or reducing major ApoA-I-binding oxidized fatty acids. Unexpectedly, oxidized fatty acid peroxisome proliferator-activated receptor γ ligands 13- and 9-hydroxyoctadecadienoic acid were increased in lymphocytes of autoimmune ApoA-I(tg) mice. ApoA-I reduced Th1 cells independently of changes in CD4(+)Foxp3(+) regulatory T cells or CD11c(+) dendritic cell activation and migration. Follicular helper T cells, germinal center B cells, and autoantibodies were also lower in ApoA-I(tg) mice. Transgenic ApoA-I also improved SLE-mediated glomerulonephritis. However, ApoA-I deficiency did not have the opposite effects on autoimmunity or glomerulonephritis, possibly as the result of compensatory increases in ApoE on high-density lipoprotein. We conclude that, although compensatory mechanisms prevent the proinflammatory effects of ApoA-I deficiency in normocholesterolemic mice, increasing ApoA-I can attenuate lymphocyte activation and autoimmunity in SLE independently of cholesterol transport, possibly through oxidized fatty acid peroxisome proliferator-activated receptor γ ligands, and it can reduce renal inflammation in glomerulonephritis.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
DBP, 98% (HPLC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)