Merck
  • Rapid Nucleation of Iron Oxide Nanoclusters in Aqueous Solution by Plasma Electrochemistry.
Rapid Nucleation of Iron Oxide Nanoclusters in Aqueous Solution by Plasma Electrochemistry.
Langmuir : the ACS journal of surfaces and colloids (2015-06-19)
Mathieu Bouchard, Mathieu Létourneau, Christian Sarra-Bournet, Myriam Laprise-Pelletier, Stéphane Turgeon, Pascale Chevallier, Jean Lagueux, Gaétan Laroche, Marc-A Fortin
ABSTRACT
Progresses in cold atmospheric plasma technologies have made possible the synthesis of nanoparticles in aqueous solutions using plasma electrochemistry principles. In this contribution, a reactor based on microhollow cathodes and operating at atmospheric pressure was developed to synthesize iron-based nanoclusters (nanoparticles). Argon plasma discharges are generated at the tip of the microhollow cathodes, which are placed near the surface of an aqueous solution containing iron salts (FeCl2 and FeCl3) and surfactants (biocompatible dextran). Upon reaction at the plasma-liquid interface, reduction processes occur and lead to the nucleation of ultrasmall iron-based nanoclusters (IONCs). The purified IONCs were investigated by XPS and FTIR, which confirmed that the nucleated clusters contain a highly hydrated form of iron oxide, close to the stoichiometric constituents of α-FeOOH (goethite) or Fe5O3(OH)9 (ferrihydrite). Relaxivity values of r1 = 0.40 mM(-1) s(-1) and r2/r1 = 1.35 were measured (at 1.41 T); these are intermediate values between the relaxometric properties of superparamagnetic iron oxide nanoparticles used in medicine (USPIO) and those of ferritin, an endogenous contrast agent. Plasma-synthesized IONCs were injected into the mouse model and provided positive vascular signal enhancement in T1-w. MRI for a period of 10-20 min. Indications of rapid and strong elimination through the urinary and gastrointestinal tracts were also found. This study is the first to report on the development of a compact reactor suitable for the synthesis of MRI iron-based contrast media solutions, on site and upon demand.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis