Merck
  • New triquinane and gymnomitrane sesquiterpenes from fermentation of the basidiomycete Antrodiella albocinnamomea.
New triquinane and gymnomitrane sesquiterpenes from fermentation of the basidiomycete Antrodiella albocinnamomea.
Fitoterapia (2015-02-24)
Zi-Ming Chen, He-Ping Chen, Fang Wang, Zheng-Hui Li, Tao Feng, Ji-Kai Liu
ABSTRACT
Five new triquinane-type sesquiterpenes, antrodins A-E (1-5) and a new gymnomitrane sesquiterpene, antrodin F (6), together with three known compounds 7-9 were isolated from fermentation of Antrodiella albocinnamomea. Their structures were elucidated through extensive spectroscopic methods including 2D NMR and HRMS analyses. The absolute configurations of 1 and 6 were determined using single-crystal X-ray diffraction analyses. The isolated compounds were investigated for their cytotoxicities against five human cancer cell lines.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis