Merck
  • Simultaneous Determination of Trigonelline, Caffeine, Chlorogenic Acid and Their Related Compounds in Instant Coffee Samples by HPLC Using an Acidic Mobile Phase Containing Octanesulfonate.
Simultaneous Determination of Trigonelline, Caffeine, Chlorogenic Acid and Their Related Compounds in Instant Coffee Samples by HPLC Using an Acidic Mobile Phase Containing Octanesulfonate.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-08-11)
Kana Arai, Hiroyuki Terashima, Sen-ichi Aizawa, Atsushi Taga, Atsushi Yamamoto, Kaname Tsutsumiuchi, Shuji Kodama
ABSTRACT
In order to analyze trigonelline, caffeine, chlorogenic acid, and their related compounds simultaneously, an HPLC method using an InertSustain C18 column and a mobile phase containing octanesulfonate as an ion-pairing reagent under an acidic condition was developed. The optimum mobile phase conditions were determined to be 0.1% phosphoric acid, 4 mM octanesulfonate, and 15% methanol at 35°C. Using the proposed method, trigonelline, nicotinic acid, caffeine, theophylline, chlorogenic acid, and caffeic acid in ten instant coffee samples were analyzed. These analytes except for theophylline were detected in all samples. An increase in the caffeine content in instant coffee samples tended to decrease in both trigonelline and chlorogenic acid contents, and the trigonelline content was found to be correlated well with the chlorogenic acid content (R(2) = 0.887).
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder