Merck
  • New abietane-type diterpene glycosides from the roots of Tripterygium wilfordii.
New abietane-type diterpene glycosides from the roots of Tripterygium wilfordii.
Journal of Asian natural products research (2015-01-16)
Hong-Mei Li, Da-Wu Wan, Rong-Tao Li
ABSTRACT
Two new abietane diterpene glycosides, wilfordosides A (1) and B (2), were isolated from the roots of Tripterygium wilfordii. The structures of compounds 1 and 2 were established using spectroscopic methods including extensive 1D and 2D NMR analysis, in combination with chemical reactions.
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
cis-4-Hydroxy-L-proline, collagen synthesis inhibitor
Sigma-Aldrich
Potassium bromide, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, powder, 99.999% trace metals basis
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
1,3-Propylene sulfite, 99%