Merck
  • High-content screening imaging and real-time cellular impedance monitoring for the assessment of chemical's bio-activation with regards hepatotoxicity.
High-content screening imaging and real-time cellular impedance monitoring for the assessment of chemical's bio-activation with regards hepatotoxicity.
Toxicology in vitro : an international journal published in association with BIBRA (2015-08-05)
Ludovic Peyre, Georges de Sousa, Sylvie Barcellini-Couget, Anne-Pascale Luzy, Nathalie Zucchini-Pascal, Roger Rahmani
ABSTRACT
Testing hepatotoxicity is a crucial step in the development and toxicological assessment of drugs and chemicals. Bio-activation can lead to the formation of metabolites which may present toxicity for the organism. Classical cytotoxic tests are not always appropriate and are often insufficient, particularly when non metabolically-competent cells are used as the model system, leading to false-positive or false-negative results. We tested over 24 h the effects of eight reference compounds on two different cell models: primary cultures of rat hepatocytes and FAO hepatoma cells that lack metabolic properties. We performed inter-assay validation between three classical cytotoxicity assays and real-time cell impedance data. We then complemented these experiments with high-content screening (HCS) to determine the cell function disorders responsible for the observed effects. Among the different assays used, the neutral red test seemed to be well suited to our two cell models, coupled with real-time cellular impedance which proved useful in the detection of bio-activation. Indeed, impedance monitoring showed a high sensitivity with interesting curve profiles yet seemed unsuitable for evaluation of viability on primary culture. Finally, HCS in the evaluation of hepatotoxicity is likely to become an essential tool for use in parallel to a classical cytotoxic assay in the assessment of drugs and environmental chemicals.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%