Merck
  • Effects of (-)-sesamin on 6-hydroxydopamine-induced neurotoxicity in PC12 cells and dopaminergic neuronal cells of Parkinson's disease rat models.
Effects of (-)-sesamin on 6-hydroxydopamine-induced neurotoxicity in PC12 cells and dopaminergic neuronal cells of Parkinson's disease rat models.
Neurochemistry international (2015-03-10)
Hyun Jin Park, Ting Ting Zhao, Kyung Sook Lee, Seung Ho Lee, Keon Sung Shin, Keun Hong Park, Hyun Sook Choi, Myung Koo Lee
ABSTRACT
The present study investigated the effects of (-)-sesamin on 6-hydroxydopamine (6-OHDA)-induced neurotoxicity using PC12 cells and dopaminergic neuronal cells of 6-OHDA-lesioned rat model of Parkinson's disease (PD). In PC12 cells, treatment with (-)-sesamin (25 µM) reduced 6-OHDA (100 µM)-induced cell death and induced transient extracellular signal-regulated kinase (ERK1/2) phosphorylation and Bad phosphorylation at Ser112 (BadSer112). In contrast, sustained ERK1/2 phosphorylation, p38 mitogen-activated protein kinase (p38MAPK) and c-Jun N-terminal kinase (JNK1/2) phosphorylation, and cleaved-caspase-3 activity, all of which were induced by 6-OHDA (100 µM), were inhibited by treatment with (-)-sesamin (25 µM). Furthermore, co-treatment with (-)-sesamin (30 mg/kg, p.o.) once a day for 28 days significantly increased the number of tyrosine hydroxylase-immunopositive neuronal cells and the levels of dopamine, norepinephrine, 3,4-dihydroxyphenylacetic acid, and homovanillic acid in the substantia nigra-striatum of 6-OHDA-lesioned rat model of PD with or without L-DOPA treatment. These results suggest that (-)-sesamin protects 6-OHDA-induced cytotoxicity via the activation of transient ERK1/2-BadSer112 system and the inhibition of sustained ERK-p38MAPK-JNK1/2-caspase-3 system in PC12 cells. (-)-Sesamin also shows protective effects on long-term L-DOPA therapy in dopaminergic neuronal cells of PD rat models. (-)-Sesamin may serve as adjuvant therapeutics in PD.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)