Merck
  • A point mutation in the acetylcholinesterase-1 gene is associated with chlorpyrifos resistance in the plant bug Apolygus lucorum.
A point mutation in the acetylcholinesterase-1 gene is associated with chlorpyrifos resistance in the plant bug Apolygus lucorum.
Insect biochemistry and molecular biology (2015-09-13)
Shuwen Wu, Kairan Zuo, Zhaokui Kang, Yihua Yang, John G Oakeshott, Yidong Wu
ABSTRACT
Control of Chinese Apolygus lucorum relies heavily on organophosphate insecticides. Here we describe resistance to the organophosphate chlorpyrifos in an A. lucorum strain, BZ-R, which was developed from a field-collected strain (BZ) by selection with chlorpyrifos in the laboratory. BZ-R showed 21-58 fold resistance to chlorpyrifos compared with the laboratory reference strain LSF and another susceptible strain, BZ-S, derived from BZ. BZ-R also showed several fold resistance to two other organophosphates and a carbamate. No synergism of chlorpyrifos by metabolic enzyme inhibitors nor any increase in detoxifying enzyme activities were observed in BZ-R. No sequence differences in acetylcholinesterase-2 were found to be associated with the resistance but the frequency of an alanine to serine substitution at position 216 of acetylcholinesterase-1 was 100% in BZ-R, ∼21-23% in SLF and BZ, and 0% in BZ-S. A single generation treatment of chlorpyrifos on the BZ strain also increased its frequency of the serine substitution to 64%. Recombinantly expressed acetylcholinesterase-1 carrying the serine substitution was about five fold less sensitive to inhibition by chlorpyrifos oxon than the wild-type enzyme. Quantitative real-time PCR found no differences in ace1 or ace2 expression levels among the strains tested. Thus the chlorpyrifos resistance is strongly associated with the serine substituted acetylcholinesterase-1. An equivalent substitution has been found to confer resistance to many organophosphate and carbamate insecticides in four other insect species.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
1-Naphthyl acetate, ≥98% (C)