Merck
  • Ultrasmall [(64)Cu]Cu nanoclusters for targeting orthotopic lung tumors using accurate positron emission tomography imaging.
Ultrasmall [(64)Cu]Cu nanoclusters for targeting orthotopic lung tumors using accurate positron emission tomography imaging.
ACS nano (2015-04-29)
Fuping Gao, Pengju Cai, Wenjiang Yang, Jingquan Xue, Liang Gao, Ru Liu, Yaling Wang, Yawei Zhao, Xiao He, Lina Zhao, Guodong Huang, Fasheng Wu, Yuliang Zhao, Zhifang Chai, Xueyun Gao
ABSTRACT
Positron emission tomography (PET) imaging has received special attention owing to its higher sensitivity, temporal resolution, and unlimited tissue penetration. The development of tracers that target specific molecules is therefore essential for the development and utility of clinically relevant PET procedures. However, (64)Cu as a PET imaging agent generally has been introduced into biomaterials through macrocyclic chelators, which may lead to the misinterpretation of PET imaging results due to the detachment and transchelation of (64)Cu. In this study, we have developed ultrasmall chelator-free radioactive [(64)Cu]Cu nanoclusters using bovine serum albumin (BSA) as a scaffold for PET imaging in an orthotopic lung cancer model. We preconjugated the tumor target peptide luteinizing hormone releasing hormone (LHRH) to BSA molecules to prepare [(64)Cu]CuNC@BSA-LHRH. The prepared [(64)Cu]Cu nanoclusters showed high radiolabeling stability, ultrasmall size, and rapid deposition and diffusion into tumor, as well as predominantly renal clearance. [(64)Cu]CuNC@BSA-LHRH showed 4 times higher tumor uptake compared with that of [(64)Cu]CuNC@BSA by analyzing the (64)Cu radioactivity of tissues via gamma counting. The PET imaging using [(64)Cu]Cu nanoclusters as tracers showed more sensitive, accurate, and deep penetration imaging of orthotopic lung cancer in vivo compared with near-infrared fluorescence imaging. The nanoclusters provide biomedical research tools for PET molecular imaging.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis