Merck
  • Quantification of dabigatran and indirect quantification of dabigatran acylglucuronides in human plasma by LC-MS/MS.
Quantification of dabigatran and indirect quantification of dabigatran acylglucuronides in human plasma by LC-MS/MS.
Bioanalysis (2015-05-13)
Shamin Mohd Saffian, Mei Zhang, Paul Ken Leong Chin, Berit Packert Jensen
ABSTRACT
An assay for the quantification of dabigatran and its active metabolites, dabigatran acylglucuronides, has not previously been described in detail. For the quantification of total dabigatran concentration (free dabigatran and acylglucuronides), samples were subjected to alkaline hydrolysis. For the quantification of free dabigatran, samples were acidified with ammonium formate. Following acetonitrile protein precipitation, the samples were analyzed by LC-MS/MS using gradient elution to ensure separation of dabigatran from dabigatran acylglucuronides. Mean recoveries ≥98% were achieved. The assay was validated over the range 2.5-1000 ng/ml dabigatran, imprecision was <9% CV (<15% at LLOQ) and accuracy was 101-114%. An assay for dabigatran with indirect quantification of dabigatran acylglucuronides in plasma was developed, validated and applied.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%