  • Search Results
  • Variable protein profiles in extracellular products of the protistan parasite Perkinsus olseni among regions of the Spanish coast.
Variable protein profiles in extracellular products of the protistan parasite Perkinsus olseni among regions of the Spanish coast.
Journal of invertebrate pathology (2015-11-12)
Sergio Fernández-Boo, Antonio Villalba, Asunción Cao
ABSTRACT
The variability of the protein expression profiling in the extracellular products (ECPs) of in vitro cultured Perkinsus olseni deriving from 4 regions of the Spanish coast was evaluated. The regions involved were the rías of Arousa and Pontevedra (Galicia, NW Spain), Carreras River (Andalusia, SW Spain) and Delta de l'Ebre (Catalonia, NE Spain). P. olseni in vitro clonal cultures were produced from parasite isolates from four clams from each region. Proteins released by the in vitro cultured parasites were isolated and separated by two dimensional electrophoresis (2DE). Qualitative comparison of protein expression profiles in the P. olseni ECPs among clones from all the regions was performed with PD Quest software. Around 130 spots were counted in the gels from ECPs of P. olseni clones from each region, of which 23 spots were shared by clones from all the regions and various spots were representative from clones of one region (appear in every clonal culture from that region but did not in every one of the other regions). A total of 34 spots were excised from the gels and analysed for sequencing. The protein cathepsin B, involved in proteolysis, the signal recognition particle receptor subunit β, involved in protein transport through membranes, and a protein belonging to N-acetyl transferase superfamily, involved in biosynthesis, were identified in spots shared by P. olseni ECPs from all regions. Pepsin A precursor, involved in proteolysis; heat shock protein (HSP) 60; and phosphoserine aminotransferase, involved in biosynthesis, were representative of P. olseni ECPs from Ría de Arousa, while peroxiredoxin V, involved in oxidation-reduction, was representative of P. olseni ECPs from Ría de Pontevedra. Differences in released proteins suggest different virulence or resistance to host attack between parasites from different locations.
Sigma-Aldrich
Chloramphenicol, γ-irradiated
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Supelco
DL-Dithiothreitol solution, 1 M in H2O