Merck
  • The viability of chondrocytes after an in vivo injection of local anaesthetic and/or corticosteroid: a laboratory study using a rat model.
The viability of chondrocytes after an in vivo injection of local anaesthetic and/or corticosteroid: a laboratory study using a rat model.
The bone & joint journal (2015-07-02)
M Sola, L Dahners, P Weinhold, A Svetkey van der Horst, S Kallianos, D Flood
ABSTRACT
This in vivo controlled laboratory study was performed to evaluate various intra-articular clinical injection regimes that might be less toxic than some in vitro studies suggest. We hypothesised that low-concentration, preservative-free, pH-balanced agents would be less toxic than high-concentration non-pH-balanced agents with preservatives, and that injections of individual agents are less toxic than combined injections. The left knees of 12- to 13-week-old Sprague-Dawley rats were injected once with eight different single agents, including low and high concentrations of ropivacaine and triamcinolone, alone and in combination, as well as negative and positive controls. The rats were killed at one week or five months, and live-dead staining was performed to quantify the death of chondrocytes. All injections except pH-balanced 0.2% ropivacaine combined with preservative-free 1 mg/ml triamcinolone acetonide resulted in statistically significant decreases in chondrocyte viability, compared with control knees, after one week and five months (p < 0.001). After one week there was no significant difference in viability between 0.2% and 0.5% ropivacaine; however, 4 mg/ml triamcinolone resulted in a lower viability than 1 mg/ml triamcinolone. Although many agents commonly injected into joints are chondrotoxic, in this in vivo study diluting preservative-free 10 mg/ml triamcinolone 1:9 in 0.2% pH-balanced ropivacaine resulted in low toxicity.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG