  • Search Results
  • Ultraviolet matrix-assisted laser desorption/ionization time-of-flight mass spectrometry for phosphopeptide analysis with a solidified ionic liquid matrix.
Ultraviolet matrix-assisted laser desorption/ionization time-of-flight mass spectrometry for phosphopeptide analysis with a solidified ionic liquid matrix.
European journal of mass spectrometry (Chichester, England) (2015-07-17)
Gargee Mukherjee, Claudia Claudia Röwer, Cornelia Koy, Chris Protzel, Peter Lorenz, Hans-Jürgen Thiesen, Oliver W Hakenberg, Michael O Glocker
ABSTRACT
A solidified ionic liquid matrix (SILM) consisting of 3-aminoquinoline, α-cyano-4- hydroxycinnamic acid and ammonium dihydrogen phosphate combines the benefits of liquid and solid MALDI matrices and proves to be well suitable for phosphopeptide analysis using MALDI-MS in the low femtomole range. Desalting and buffer exchange that typically follow after phosphopeptide elution from metal oxide affinity chromatography (MOAC) materials can be omitted. Shifting the pH from acidic to basic during target preparation causes slow matrix crystallization and homogeneous embedding of the analyte molecules, forming a uniform preparation from which (phospho)peptides can be ionized in high yields over long periods of time. The novel combination of MOAC-based phosphopeptide enrichment with SILM preparation has been developed with commercially available standard phosphopeptides and with α-casein as phosphorylated standard protein. The applicability of the streamlined phosphopeptide analysis procedure to cell biological and clinical samples has been tested (i) using affinity-enriched endogenous TRIM28 from cell cultures and (ii) by analysis of a two-dimensional gel-separated protein spot from a bladder cancer sample.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O