Merck
  • Characterization of Staphylococcus aureus Strains Isolated from Czech Cystic Fibrosis Patients: High Rate of Ribosomal Mutation Conferring Resistance to MLS(B) Antibiotics as a Result of Long-Term and Low-Dose Azithromycin Treatment.
Characterization of Staphylococcus aureus Strains Isolated from Czech Cystic Fibrosis Patients: High Rate of Ribosomal Mutation Conferring Resistance to MLS(B) Antibiotics as a Result of Long-Term and Low-Dose Azithromycin Treatment.
Microbial drug resistance (Larchmont, N.Y.) (2015-04-01)
Jan Tkadlec, Eva Vařeková, Roman Pantůček, Jiří Doškař, Vladislava Růžičková, Tibor Botka, Libor Fila, Oto Melter
ABSTRACT
Staphylococcus aureus is one of the most frequent pathogens infecting the respiratory tract of patients with cystic fibrosis (CF). This study was the first to examine S. aureus isolates from CF patients in the Czech Republic. Among 100 S. aureus isolates from 92 of 107 observed patients, we found a high prevalence of resistance to macrolide-lincosamide-streptogramin B (MLS(B)) antibiotics (56%). More than half of the resistant strains (29 of 56) carried a mutation in the MLS(B) target site. The emergence of MLS(B) resistance and mutations conferring resistance to MLS(B) antibiotics was associated with azithromycin treatment (p=0.000000184 and p=0.000681, respectively). Methicillin resistance was only detected in 3% of isolates and the rate of resistance to other antibiotics did not exceed 12%. The prevalence of small-colony variant (SCV) strains was relatively low (9%) and eight of nine isolates with the SCV phenotype were thymidine dependent. The study population of S. aureus was heterogeneous in structure and both the most prevalent community-associated and hospital-acquired clonal lineages were represented. Of the virulence genes, enterotoxin genes seg (n=52), sei (n=49), and sec (n=16) were the most frequently detected among the isolates. The PVL genes (lukS-PV and lukF-PV) have not been revealed in any of the isolates.
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications