Merck
  • Increased levels of galectin-3 were associated with prediabetes and diabetes: new risk factor?
Increased levels of galectin-3 were associated with prediabetes and diabetes: new risk factor?
Journal of endocrinological investigation (2014-12-17)
H Yilmaz, M Cakmak, O Inan, T Darcin, A Akcay
ABSTRACT
Galectin-3 (Gal-3) is a marker of cardiac fibrosis and predicts incident heart failure. Gal-3-deficient mice are resistant to multiple low-dose streptozotocin-induced diabetes. Recent experimental studies suggested an important role for Gal-3 in the regulation of adiposity, metaflammation and type 2 diabetes. This study aimed to examine the relationship between Gal-3 and newly diagnosed prediabetes and diabetes. Gal-3 concentrations were measured in 118 participants and 56 age- and sex-matched healthy controls. All subjects underwent a 75-g oral glucose tolerance test and were stratified into normal, prediabetic, and diabetes mellitus subgroups. DM was defined as a plasma glucose level ≥126 mg/dL in the fasting state or ≥200 mg/dL in the second hour after glucose loading. Impaired fasting glucose was defined as an FPG level of 100-125 mg/dL, and impaired glucose tolerance was defined as a 2-h plasma glucose level of 140-199 mg/dL. Sixty-one patients had prediabetes (Group 1), 57 had diabetes (Group 2), and 56 had neither diabetes nor prediabetes (Group 3). Gal-3 levels correlated with FPG (r = 0.787, P < 0.01), 2hPG (r = 0.833, P < 0.01), CRP (r = 0.501, P < 0.01), and HOMA-IR (r = 0.518, P < 0.01). Gal-3 levels were higher in Group 2 than in Groups 1 and 3 [1,053.9 (358.1) and 744.1 (119.3) vs. 481.7 (175.4) pg/mL; P < 0.001]. Gal-3 is an independent predictor of diabetes in multivariate logistic analysis. In ROC analysis, a Gal-3 cutoff value of 803.55 pg/mL diagnoses diabetes with a sensitivity of 80.7 % and a specificity of 85.5 % (AUC = 0.912). Gal-3 is a promising biomarker for detecting prediabetes and diabetes.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline