Merck
  • Useful properties of undifferentiated mesenchymal stromal cells and adipose tissue as the source in liver-regenerative therapy studied in an animal model of severe acute fulminant hepatitis.
Useful properties of undifferentiated mesenchymal stromal cells and adipose tissue as the source in liver-regenerative therapy studied in an animal model of severe acute fulminant hepatitis.
Cytotherapy (2015-07-04)
Bruna Maria Manzini, Adriana da Silva Santos Duarte, Sundararaj Sankaramanivel, Aline Lisie Ramos, Paulo Latuf-Filho, Cecilia Escanhoela, Paulo Kharmandayan, Sara Teresinha Olalla Saad, Ilka Boin, Ângela Cristina Malheiros Luzo
ABSTRACT
End-stage liver diseases frequently require liver transplantation. Cell therapy could be an alternative. This study aimed to analyze whether undifferentiated mesenchymal stromal cells (U-MSCs) or MSC-derived hepatocyte-like cells (DHLCs) from adipose tissue (AT), umbilical cord blood (UCB) and bone marrow (BM) would better restore damaged liver. AT was obtained from lipo-aspiration, UCB from an Umbilical Cord Blood Bank and BM from a BM Transplantation Unit. AT (collagenase digestion), UCB and BM (Ficoll gradient) were cultured (Dulbecco's modified Eagle's medium, low glucose, FBS) for 3 days. Detached adherent cells, at passage 4, were characterized as MSCs. Genetic stability was investigated by means of telomerase enzyme activity and karyotype. Hepatocyte differentiation protocol was performed with the use of Dulbecco's modified Eagle's medium, hepatocyte growth factor, basic fibroblast growth factor and nicotinamide (7 days); maturation medium (oncostatin, dexamethasone, insulin, transferrin and selenium) was added at 36 days. Hepatogenesis analyses were performed by use of morphology and albumin, AF, tyrosine-aminotransferase and glutamine synthetase gene expression and quantitative reverse transcription-polymerase chain reaction on days 9, 18, 25 and 36. Functionality was assessed through glycogen storage detection, indocyanine green absorption and transplantation procedure. U-MSCs and DHLCs were injected 48 h after induced fulminant hepatitis (intraperitoneal injection of carbon tetrachloride) in SCID/BALB-c mice. Histopathologic analyses were performed on days 7 and 15. Human origin included albumin and CK19 human markers. All MSCs differentiated into functional hepatocyte-like cells, stored glycogen and absorbed indocyanine green. AT-MSC DHLC gene expression was more consistent with a normal hepatogenic-differentiation profile. UCB-MSCs expanded weakly, impairing their use for the transplantation procedure. AT and BM U-MSCs and DHLCs regenerated liver injury equally. Regenerated hepatocytes exhibited human origin. AT might be the source and U-MSCS the stem cells useful for liver-regenerative therapy.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder