Merck
  • Chromatographic efficiency comparison of polyhedral oligomeric silsesquioxanes-containing hybrid monoliths via photo- and thermally-initiated free-radical polymerization in capillary liquid chromatography for small molecules.
Chromatographic efficiency comparison of polyhedral oligomeric silsesquioxanes-containing hybrid monoliths via photo- and thermally-initiated free-radical polymerization in capillary liquid chromatography for small molecules.
Journal of chromatography. A (2015-08-04)
Hongwei Wang, Junjie Ou, Zhongshan Liu, Hui Lin, Xiaojun Peng, Hanfa Zou
ABSTRACT
Monolithic poly(methacrylate epoxy cyclosiloxane-co-polyhedral oligomeric silsesquioxanes) (epoxy-MA-POSS) capillary columns have been prepared via either photo- or thermally-initiated polymerization of the corresponding monomers using a 1-propanol/PEG 400 mixture as porogens. Photochemical polymerization was accomplished by irradiation of the UV-transparent capillary for 10min at room temperature, while thermal polymerization was performed at 55°C, 60°C or 65°C for 18h. The evaluation of chromatographic property for two hybrid epoxy-MA-POSS monoliths was carried out. The results indicate that hybrid monoliths fabricated by photochemical initiation exhibit higher column efficiency (97,000-98,400plates/m) than those synthesized by thermal polymerization (41,100-48,000plates/m) in cLC. The higher efficiency of photo-initiated hybrid monoliths is closely related to lower eddy dispersion (A-term) and mass transfer resistance (C-term).
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%