Merck
  • Functional Dopaminergic Neurons in Substantia Nigra are Required for Transcranial Magnetic Stimulation-Induced Motor Plasticity.
Functional Dopaminergic Neurons in Substantia Nigra are Required for Transcranial Magnetic Stimulation-Induced Motor Plasticity.
Cerebral cortex (New York, N.Y. : 1991) (2014-01-24)
Tsung-Hsun Hsieh, Ying-Zu Huang, Alexander Rotenberg, Alvaro Pascual-Leone, Yung-Hsiao Chiang, Jia-Yi Wang, Jia-Jin J Chen
ABSTRACT
Repetitive magnetic stimulation (rTMS), including theta burst stimulation (TBS), is capable of modulating motor cortical excitability through plasticity-like mechanisms and might have therapeutic potential for Parkinson's disease (PD). An animal model would be helpful for elucidating the mechanism of rTMS that remain unclear and controversial. Here, we have established a TMS model in rat and applied this model to study the impact of substantia nigra dopamine neuron on TBS-induced motor plasticity in PD rats. In parallel with human results, continuous TBS (cTBS) successfully suppressed motor evoked potentials (MEPs), while MEPs increased after intermittent TBS (iTBS) in healthy rats. We then tested the effect of iTBS in early and advanced 6-hydroxydopamine (6-OHDA)-lesioned PD. Moreover, dopaminergic neurons in substantia nigra and rotation behavior were assessed to correlate with the amount of iTBS-induced plasticity. In results, iTBS-induced potentiation was reduced in early PD rats and was absent in advanced PD rats. Such reduction in plasticity strongly correlated with the dopaminergic cell loss and the count of rotation in PD rats. In conclusion, we have established a TMS PD rat model. With the help of this model, we confirmed the loss of domaninergic neurons in substantia nigra resulting in reduced rTMS-induced motor plasticity in PD.
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder