Merck
  • Antioxidant activity of polysaccharide purified from Acanthopanax koreanum Nakai stems in vitro and in vivo zebrafish model.
Antioxidant activity of polysaccharide purified from Acanthopanax koreanum Nakai stems in vitro and in vivo zebrafish model.
Carbohydrate polymers (2015-05-13)
Min-Cheol Kang, Seo-Young Kim, Eun-A Kim, Ji-Hyeok Lee, Young-Sun Kim, Seok-Kyu Yu, Jun Byoung Chae, In-Hu Choe, Jin Hun Cho, You-Jin Jeon
ABSTRACT
In this study, the antioxidative effects of a purified polysaccharide isolated from the stems of Acanthopanax koreanum Nakai (ASP) on hydrogen peroxide-induced oxidative stress was investigated both in vitro and in vivo using a zebrafish model. A. koreanum Nakai stem was hydrolyzed using five carbohydrases and five proteases for the enzyme-assistant extraction. Of the enzyme-assistant extracts, the Protamex extract exhibited the highest yield and a potent scavenging activity against free radicals. Ethanol-added separation and anion exchange chromatography were conducted to identify the active polysaccharide. The purified polysaccharide significantly scavenged hydrogen peroxide and reduced hydrogen peroxide-induced cell death in Vero cells and in zebrafish. The results reveal that ASP is a useful antioxidant polysaccharide and might be available for relevant industrial applications.
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)