  • Search Results
  • The small GTPase Rab29 is a common regulator of immune synapse assembly and ciliogenesis.
The small GTPase Rab29 is a common regulator of immune synapse assembly and ciliogenesis.
Cell death and differentiation (2015-05-30)
A Onnis, F Finetti, L Patrussi, M Gottardo, C Cassioli, S Spanò, C T Baldari
ABSTRACT
Accumulating evidence underscores the T-cell immune synapse (IS) as a site of intense vesicular trafficking, on which productive signaling and cell activation crucially depend. Although the T-cell antigen receptor (TCR) is known to exploit recycling to accumulate to the IS, the specific pathway that controls this process remains to be elucidated. Here we demonstrate that the small GTPase Rab29 is centrally implicated in TCR trafficking and IS assembly. Rab29 colocalized and interacted with Rab8, Rab11 and IFT20, a component of the intraflagellar transport system that regulates ciliogenesis and participates in TCR recycling in the non-ciliated T cell, as assessed by co-immunoprecipitation and immunofluorescence analysis. Rab29 depletion resulted in the inability of TCRs to undergo recycling to the IS, thereby compromizing IS assembly. Under these conditions, recycling TCRs accumulated in Rab11(+) endosomes that failed to polarize to the IS due to defective Rab29-dependent recruitment of the dynein microtubule motor. Remarkably, Rab29 participates in a similar pathway in ciliated cells to promote primary cilium growth and ciliary localization of Smoothened. These results provide a function for Rab29 as a regulator of receptor recycling and identify this GTPase as a shared participant in IS and primary cilium assembly.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
MISSION® esiRNA, targeting RLUC
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)