Merck
  • Transition metal selenides as efficient counter-electrode materials for dye-sensitized solar cells.
Transition metal selenides as efficient counter-electrode materials for dye-sensitized solar cells.
Physical chemistry chemical physics : PCCP (2015-10-13)
Jiahao Guo, Suxia Liang, Yantao Shi, Ce Hao, Xuchun Wang, Tingli Ma
ABSTRACT
Exploiting an alternative of the Pt-based counter-electrode materials for the triiodide reduction reaction has become a major interest in the fundamental research of dye-sensitized solar cells. Transition-metal selenides have recently been demonstrated as promising non-precious metal electrocatalysts for the triiodide reduction reaction. Herein, we prepared a series of transition-metal selenides via a free-reductant solvothermal method and used them as counter-electrodes in high efficiency dye-sensitized solar cells. The electrochemical results showed that these selenides had excellent catalytic activity for the reduction of the triiodine/iodine couple, and except for MoSe2, the conversion efficiencies of the corresponding dye-sensitized solar cells were comparable to the sputtered Pt counter-electrode. Theoretical investigation clearly revealed that the unsatisfactory performance of MoSe2 mainly originated from the processes of adsorption and charge-transfer. These findings can help to better understand the electrocatalytic processes and thus offer some useful guidelines to develop more efficient electrochemical catalysts.
Sigma-Aldrich
Lithium diisopropylamide, 97%
Sigma-Aldrich
Lithium diisopropylamide solution, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Lithium diisopropylamide solution, 1.0 M in THF/hexanes
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)