Merck
  • A reversed-phase/hydrophilic interaction mixed-mode C18-Diol stationary phase for multiple applications.
A reversed-phase/hydrophilic interaction mixed-mode C18-Diol stationary phase for multiple applications.
Analytica chimica acta (2015-09-01)
Qing Wang, Mao Ye, Li Xu, Zhi-guo Shi
ABSTRACT
A mixed-mode chromatographic packing material, C18 and diol groups modified silica (C18-Diol), was prepared with controllable hydrophobicity and hydrophilicity. It demonstrated excellent aqueous compatibility and stability in aqueous mobile phase; compared to the traditional C18 column, improved peak shape of basic analytes was also obtained. Additionally, it exhibited both reversed-phase liquid chromatographic (RPLC) and hydrophilic interaction chromatographic (HILIC) performance; the analyte separation scope was thus enlarged, demonstrated by simultaneous separation of twenty acids, bases and neutrals. More interestingly, a novel on-line two-dimensional liquid chromatography on the single column (2D-LC-1C) was established by modifying the high performance liquid chromatographic instrument only with the addition of an extra six-port two-position valve. The early co-eluted components of the extract of Lonicera japonica on the 1st-dimension (RPLC) were collected for the online re-injection to the 2nd-dimension (HILIC) by conveniently varying the mobile phase components. Six more peaks were obtained. The established system was simple, easy operation and low cost, which had advantages in analyzing complicated samples.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Tetracaine, ≥98% (TLC)
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Cytosine, ≥99%
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)