Merck
  • Enhancing the vitamin B12 production and growth of Propionibacterium freudenreichii in tofu wastewater via a light-induced vitamin B12 riboswitch.
Enhancing the vitamin B12 production and growth of Propionibacterium freudenreichii in tofu wastewater via a light-induced vitamin B12 riboswitch.
Applied microbiology and biotechnology (2015-09-17)
Yue Yu, Xuan Zhu, Yubiao Shen, Huanghong Yao, Peiheng Wang, Kun Ye, Xiaofeng Wang, Qing Gu
ABSTRACT
The vitamin B12-dependent riboswitch is a crucial factor that regulates gene transcription to mediate the growth of and vitamin B12 synthesis by Propionibacterium freudenreichii. In this study, the effect of various wavelengths of light on the growth rate and vitamin B12 synthesis was studied. Red, green, and blue light-emitting diodes (LEDs) were selected, and a dark condition was used as the control. The microorganism growth rate was measured using a spectrophotometer and plate counting, while the vitamin B12 content was determined using an HPLC-based method. The optical density at 600 nm (OD600) values indicated that P. freudenreichii grew better under the continuous and discontinuous blue light conditions. Moreover, under the blue light condition, P. freudenreichii tended to have a higher growth rate (0.332 h(-1)) and vitamin B12 synthesis (ca. 10 μg/mL) in tofu wastewater than in dark conditions. HPLC analysis also showed that more methylcobalamin was produced under the blue light conditions than in the other conditions. The cbiB gene transcription results showed that blue light induced the synthesis of this vitamin B12 synthesis enzyme. Moreover, the results of inhibiting the expression of green fluorescent protein indicated that blue light removed the inhibition by the vitamin B12-dependent riboswitch. This method can be used to reduce fermentation time and produce more vitamin B12 in tofu wastewater.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%