Merck
  • The Bangladesh Risk of Acute Vascular Events (BRAVE) Study: objectives and design.
The Bangladesh Risk of Acute Vascular Events (BRAVE) Study: objectives and design.
European journal of epidemiology (2015-05-02)
Rajiv Chowdhury, Dewan S Alam, Ismail Ibrahim Fakir, Sheikh Daud Adnan, Aliya Naheed, Ishrat Tasmin, Md Mostafa Monower, Farzana Hossain, Fatema Mahjabin Hossain, Md Mostafizur Rahman, Sadia Afrin, Anjan Kumar Roy, Minara Akter, Sima Akter Sume, Ajoy Kumer Biswas, Lisa Pennells, Praveen Surendran, Robin D Young, Sarah A Spackman, Khaled Hasan, Eric Harshfield, Nasir Sheikh, Richard Houghton, Danish Saleheen, Joanna M M Howson, Adam S Butterworth, Rubhana Raqib, Abdulla Al Shafi Majumder, John Danesh, Emanuele Di Angelantonio
ABSTRACT
During recent decades, Bangladesh has experienced a rapid epidemiological transition from communicable to non-communicable diseases. Coronary heart disease (CHD), with myocardial infarction (MI) as its main manifestation, is a major cause of death in the country. However, there is limited reliable evidence about its determinants in this population. The Bangladesh Risk of Acute Vascular Events (BRAVE) study is an epidemiological bioresource established to examine environmental, genetic, lifestyle and biochemical determinants of CHD among the Bangladeshi population. By early 2015, the ongoing BRAVE study had recruited over 5000 confirmed first-ever MI cases, and over 5000 controls "frequency-matched" by age and sex. For each participant, information has been recorded on demographic factors, lifestyle, socioeconomic, clinical, and anthropometric characteristics. A 12-lead electrocardiogram has been recorded. Biological samples have been collected and stored, including extracted DNA, plasma, serum and whole blood. Additionally, for the 3000 cases and 3000 controls initially recruited, genotyping has been done using the CardioMetabochip+ and the Exome+ arrays. The mean age (standard deviation) of MI cases is 53 (10) years, with 88 % of cases being male and 46 % aged 50 years or younger. The median interval between reported onset of symptoms and hospital admission is 5 h. Initial analyses indicate that Bangladeshis are genetically distinct from major non-South Asian ethnicities, as well as distinct from other South Asian ethnicities. The BRAVE study is well-placed to serve as a powerful resource to investigate current and future hypotheses relating to environmental, biochemical and genetic causes of CHD in an important but under-studied South Asian population.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications