Merck
  • Urinary metabonomics elucidate the therapeutic mechanism of Orthosiphon stamineus in mouse crystal-induced kidney injury.
Urinary metabonomics elucidate the therapeutic mechanism of Orthosiphon stamineus in mouse crystal-induced kidney injury.
Journal of ethnopharmacology (2015-03-22)
Songyan Gao, Wei Chen, Zhongjiang Peng, Na Li, Li Su, Diya Lv, Ling Li, Qishan Lin, Xin Dong, Zhiyong Guo, Ziyang Lou
ABSTRACT
Orthosiphon stamineus (OS), a traditional Chinese herb, is often used for promoting urination and treating nephrolithiasis. Urolithiasis is a major worldwide public health burden due to its high incidence of recurrence and damage to renal function. However, the etiology for urolithiasis is not well understood. Metabonomics, the systematic study of small molecule metabolites present in biological samples, has become a valid and powerful tool for understanding disease phenotypes. In this study, a urinary metabolic profiling analysis was performed in a mouse model of renal calcium oxalate crystal deposition to identify potential biomarkers for crystal-induced renal damage and the anti-crystal mechanism of OS. Thirty six mice were randomly divided into six groups including Saline, Crystal, Cystone and OS at dosages of 0.5g/kg, 1g/kg, and 2g/kg. A metabonomics approach using ultra-performance liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry (UHPLC-Q-TOF/MS) was developed to perform the urinary metabolic profiling analysis. Principal component analysis (PCA) and partial least squares discriminant analysis (PLS-DA) were utilized to identify differences between the metabolic profiles of mice in the saline control group and crystal group. Using partial least squares-discriminant analysis, 30 metabolites were identified as potential biomarkers of crystal-induced renal damage. Most of them were primarily involved in amino acid metabolism, taurine and hypotaurine metabolism, purine metabolism, and the citrate cycle (TCA). After the treatment with OS, the levels of 20 biomarkers had returned to the levels of the control samples. Our results suggest that OS has a protective effect for mice with crystal-induced kidney injury via the regulation of multiple metabolic pathways primarily involving amino acid, energy and choline metabolism.
Sigma-Aldrich
L-Tyrosine, FG
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
SAFC
Taurine
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis